Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(I) iodide, ultra dry, 99.998% (metals basis)

Catalog No. AA3570006
Click to view available options
Quantity:
1 g
5 g

CAS: 13966-94-4

Indium(I) iodide can be used to prepare indium cluster and chain compounds. It is also used as a reagent to propagate organic reactions viz. cyclization and cleavage of diselenide or disulfide. It plays an important role in preparing indium containing complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13966-94-4
MDL Number MFCD00135543

Specifications

Melting Point 351°C
Physical Form Beads
Quantity 5 g
Solubility Information Slightly soluble in some organic solvents,pyridine or m-xylene mixture.
Formula Weight 241.72
Percent Purity 99.998%
Packaging Ampule
Sensitivity Light and moisture sensitive
Assay (metals basis)
Chemical Name or Material Indium(I) iodide, Ultra dry
Hazard Category H302-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
EINECSNumber 237-746-3
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.