Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) acetate, 99.99% (metals basis)

Catalog No. aa4223009
Click to view available options
Quantity:
10 g
50 g

CAS: 25114-58-3 | C2H4InO2 | 174.87 g/mol

Indium(III) acetate is used as a catalyst in intermolecular radical addition of organic iodides to electron deficient alkenes. It is also used in the reduction of 1-bromo-3-phenylpropane to propylbenzene. Further, it is used to prepare the indium oxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25114-58-3
Molecular Formula C2H4InO2
Molecular Weight (g/mol) 174.87
MDL Number MFCD00054347
InChI Key LGHOYKSQIQISBI-UHFFFAOYSA-N
Synonym indium acetate, acetic acid, indium 3+ salt
PubChem CID 24191761
IUPAC Name acetic acid;indium
SMILES CC(=O)O.[In]

Specifications

Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99% (metals basis)
Solubility Information Soluble in water,mineral acid and acetic acid.
Formula Weight 291.96
Sensitivity Hygroscopic
Chemical Name or Material Indium(III) acetate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.