Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) hydroxide, 99.8% (metals basis)

Catalog No. p-7050962
Click to view available options
Quantity:
10 g
50 g

CAS: 20661-21-6

Indium(III) hydroxide is used as a precursor to indium(III)oxide. It is also applied in batteries, glasses, ceramics, chemical reagents and semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20661-21-6
MDL Number MFCD00011056

Specifications

Melting Point 150°C (in -H2O)
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 165.84
Percent Purity 99.8%
Assay (metals basis)
Chemical Name or Material Indium(III) hydroxide
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 243-947-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.