Learn More
Indium(III) oxide, 99.99% (metals basis)
Description
Indium oxide is widely utilized as a n-type semiconductor, which is used in integrated circuits as a resistive element. It finds applications in batteries and as a part of some stain formulations. It is used as thin film coatings in optical, antistatic and infrared reflectors. It is doped with tin oxide and used as transparent conductive coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 850° volat. |
Color | Yellow |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.994% (Metal basis) |
Solubility Information | Insoluble in water and soluble in acids. |
Formula Weight | 277.64 |
Odor | Odorless |
Appearance | Yellow powder/granules |
Density | 7.18 |
Show More |
Safety and Handling
EINECSNumber | 215-193-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.