Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) selenide, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8828006

Quantity 5g

 View more versions of this product

Catalog No. AA8828006


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12056-07-4
99.99% (Metal basis)
466.52
Indium(III) selenide
3 to 6mm Pieces
In2Se3
MFCD00016147

Specifications

12056-07-4
99.99% (Metal basis)
Silver gray pieces
MFCD00016147
Odorless
5g
3 to 6mm Pieces
In2Se3
466.52
5.67g/cm3
Silver-gray
Indium(III) selenide

Safety and Handling

SDS & Safety Documents

H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

EINECSNumber : 235-016-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit