Alfa Aesar™ Indium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1071109

Quantity 10g

 View more versions of this product

Catalog No. AA1071109

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 13464-82-9
    Formula Weight 517.83 (Anhydrous)
    MDL Number MFCD00149747
    Chemical Name or Material Indium(III) sulfate hydrate
    Grade Puratronic™

    Specifications

    Formula Weight 517.83 (Anhydrous)
    Physical Form Lump/Powder
    MDL Number MFCD00149747
    Quantity 10g
    TSCA TSCA
    CAS 13464-82-9
    Melting Point 250°C (dec.)
    Chemical Name or Material Indium(III) sulfate hydrate
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit