Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indium(III) sulfide, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Indium(III) sulfide emerged as a promising low-hazard buffer material and is used to improve the properties of solar cells and also replaces the hazardous CdS. It is a mid band gap semiconductor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1050°C |
Color | Red to Orange |
Physical Form | Powder |
Quantity | 50 g |
Solubility Information | Insoluble in water and organic solvents. |
Formula Weight | 325.83 |
Percent Purity | 99.995% |
Odor | Like rotten eggs |
Sensitivity | Moisture sensitive |
Assay | (metals basis) |
Show More |
Safety and Handling
Hazard Category | H302+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H332-H315-H319-H335 Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 234-742-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction