Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) trifluoromethanesulfonate, 99% min

Catalog No. p-7050971
Click to view available options
Quantity:
10 g
50 g

Catalog No. AA4013109

Supplier: Thermo Scientific Chemicals 04013109

Only null left
Add to Cart

CAS: 128008-30-0 | C3F9InO9S3 | 562.008 g/mol

Indium(III) trifluoromethanesulfonate is used as a Lewis acid catalyst in organic synthesis and as a co-catalyst in organometallic catalysis. It is also used as a reactant in the preparation of stable indium bacteriochlorins and decahydroquinoline-type toxins through intramolecular hetero Diels-Alder reactions. Further, it is used in the synthesis of benzoxazoles through cyclocondensations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128008-30-0
Molecular Formula C3F9InO9S3
Molecular Weight (g/mol) 562.008
MDL Number MFCD00144478
InChI Key UCYRAEIHXSVXPV-UHFFFAOYSA-K
Synonym indium iii trifluoromethanesulfonate, indium triflate, indium trifluoromethanesulphonate, indium 3+ tritriflate, indium 3+ ; trifluoromethanesulfonate, indium iii triflate, indium iii trifluoromethanesulphonate, acmc-209bda, in tfa 3, ksc491q8t
PubChem CID 2734680
IUPAC Name indium(3+);trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3]

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 10 g
UN Number UN3261
Solubility Information Slightly soluble in hydrocarbons and diethyl ether. Insoluble in water
Formula Weight 562.02
Percent Purity ≤99%
Sensitivity Hygroscopic
Chemical Name or Material Indium(III) trifluoromethanesulfonate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.