Alfa Aesar™ Indium shot, tear drop, 4mm (0.2 in.), 99.9% (metals basis)

Manufacturer: Alfa Aesar™ 1102630

Quantity 250g

 View more versions of this product

Catalog No. AA1102630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-74-6
    MDL Number MFCD00134048
    Chemical Name or Material Indium shot, 4mm (0.2 in.)

    Specifications

    Physical Form Teardrop Shot
    MDL Number MFCD00134048
    CAS 7440-74-6
    Odor Odorless
    TSCA TSCA
    Quantity 250g
    Chemical Name or Material Indium shot, 4mm (0.2 in.)
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit