Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Individual Instruction: Introduction To Mendelian Genetics
SCI_ED

Catalog No. S01995 Shop All Aldon Products
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S01995 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01995 Supplier Innovating Science™ Supplier No. IS1521
Add to Cart
Add to Cart

In this activity, students can simulate Mendel's work and determine patterns of inheritance using special chips and Innovating Science's exclusive 'double dice.'

In this activity, students can simulate Mendel's work and determine patterns of inheritance. Using special chips and Innovating Science's exclusive 'double dice,' students will be able to simulate both monohybrid and dihybrid crosses. After the crosses, students will be able determine genotypic and phenotypic ratios for select traits and compare their values to the theoretical 'ideal' values as put forth by Mendel. Kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.

Specifications

Grade 6+
Class Size 1 Lab Group
Product Type Genes and Probability Genetic Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.