Description
- To prevent any damage to the surface of weights, we offer you a wide range of accessories for the proper handling, storage and cleaning of weights.
Specifications
Specifications
| Product Type | Calibration Mass |
| Weight (English) | 0.02205 Ib. |
| Classification | ASTM Class 1 |
| Material | Stainless Steel |
| Weight (Metric) | 10 g |
| Tolerance | 0.002 to 0.004 |