Learn More
Sartorius Individual Precise Calibration Weight
Catalog No. 14558774 Shop All Sartorius Products
Change view
Click to view available options
Weight (Metric):
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Weight (Metric)
14-558-774 2 g
14-558-753 500 mg
14-558-754 200 mg
14-558-755 100 mg
14-558-756 50 mg
14-558-757 20 mg
14-558-758 10 mg
14-558-759 5 mg
14-558-760 2 mg
14-558-761 1 mg
14-558-763 10 kg
14-558-764 5 kg
14-558-765 2 kg
14-558-766 1 kg
14-558-767 500 g
14-558-768 200 g
14-558-769 100 g
14-558-770 50 g
14-558-771 20 g
14-558-772 10 g
14-558-773 5 g
14-558-775 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 14-558-774 Supplier Sartorius Supplier No. WI002GAC1N
Add to Cart
Add to Cart
ASTM Class 1 Individual Precise Calibration Weight without Certificate.
  • To prevent any damage to the surface of weights, we offer you a wide range of accessories for the proper handling, storage and cleaning of weights.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Mass
Weight (English) 0.0044 Ib.
Classification ASTM Class 1
Material Stainless Steel
Weight (Metric) 2 g
Tolerance 0.002 to 0.004