Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Indo-1, AM, cell permeant - Special Packaging

Catalog No. I1223
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 x 50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I1223 20 x 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I1223 Supplier Molecular Probes™ Supplier No. I1223
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Indo-1, AM, cell permeant - Special Packaging

This cell-permeant indo-1, AM is a UV light—excitable, ratiometric Ca2+ indicator. The emission maximum of indo-1 shifts from ∼475 nm in Ca2+-free medium to ∼400 nm when the dye is saturated with Ca2+. This AM ester form of the dye is available in 1 mg amounts (I-1203) and in 1 mM solution (I-1226). A cell-impermeant pentapotassium (F-1202) salt form of the dye is also available.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer -5°C to -30°C and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Indicator, Flow Cytometry
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Indo-1 AM
Dye Type Indo-1
Excitation Wavelength Range 330 to 345 nm
Form Solid
Quantity 20 x 50 μg
Shipping Condition Room Temperature
Emission Wavelength Range 390 to 405 nm
Excitation Wavelength 330 nm
Emission Wavelength 401 nm
Show More Show Less
I am doing calcium flux imaging with your Fura-2 calibration kit, but am seeing a large variability in ratio in different places around the slide. I am correcting for uniform illumination, using the product as directed, and sealing the coverslip with nail polish.

The nail polish may be the problem. The Kd value (calcium sensitivity) changes depending upon the dye's environment. Nail polish has solvents that can leech under the coverslip and cause variability. We recommend either going without a sealing or sealing with melted paraffin painted on the coverslip edges with a cotton-tipped applicator (paraffin is hydrophobic and has no solvents).

I need to label cells with Fluo-4, AM, for a calcium flux assay. How long after labeling will the dye be retained?

After loading dye into the cells, intracellular esterases remove the 'AM' moiety from the dye. When the 'AM' group is removed, the dye is able to bind calcium and fluoresce. Since the dye is not covalently bound to any cellular components, it may be actively effluxed from the cell. The rate of efflux is dependent upon the inherent properties of the cell, culture conditions and other factors. The dye may be retained for hours, days or even weeks or lost in a matter of minutes. The use of Probenecid (Cat. No. P36400) limits loss by active efflux.

What indicator dye can I use to measure calcium flux by flow cytometry?

Indo-1, AM is the preferred stain for flow cytometry, where it is more practical to use a single laser for excitation, usually the 351–364 nm spectral lines of the argon-ion laser, and monitor two emissions. The emission maximum of Indo-1 shifts from ~475 nm in Ca2+-free medium to ~400 nm when the dye is saturated with Ca2+. Indo-1, AM is particularly suited for multicolor fluorescence applications.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.