Learn More
Description
This product is supplied with a high degree of hydration and may contain residual inorganic salts, the amount of which are batch dependent. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.
Specifications
Specifications
| Chemical Name or Material | Sodium 4-(2-((1 E,3 E,5 E,7 Z)-7-(1,1-Dimethyl-3-(4-sulfonatobutyl)-1,3-dihydro-2 H-benzo[e]indol-2-ylidene)hepta-1,3,5-trien-1-yl)-1,1-dimethyl-1 H-benzo[e]indol-3-ium-3-yl)butane-1-sulfonate |
| CAS | 3599-32-4 |
| Quantity | 10 mg |
| Product Line | Tocris |
| Target | Fluorescent Dyes |
| Molecular Formula | C43H47N2O6S2Na |
| Purity | 95% |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
