Learn More
Indole-2-carboxylic acid, 98+%
Description
Reactant for total synthesis of (±)-dibromophakellin and analogs, synthesis of the pyrrolizidine alkaloid (±)-trachelanthamidine, stereo selective preparation of renieramycin G analogs, preparation of spirooxoindolepyrrolidines via reduction of indole-2-carboxylic acid followed by oxidation, condensation, reduction, amidation and Kharasch radical cyclization, Pd-catalyzed cyclization and N,N-(pentane)diylbis[indolecarboxamide] and N,N-[phenylenebis(methylene)]bis[indolecarboxamide] derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reactant for total synthesis of (±)-dibromophakellin and analogs, synthesis of the pyrrolizidine alkaloid (±)-trachelanthamidine, stereo selective preparation of renieramycin G analogs, preparation of spirooxoindolepyrrolidines via reduction of indole-2-carboxylic acid followed by oxidation, condensation, reduction, amidation and Kharasch radical cyclization, Pd-catalyzed cyclization and N,N′-(pentane)diylbis[indolecarboxamide] and N,N′-[phenylenebis(methylene)]bis[indolecarboxamide] derivatives.
Solubility
Soluble in clear yellow solution 5% in ethanol, dimethyl sulfoxide and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | ∼205°C (decomposition) |
| Quantity | 10 g |
| Beilstein | 124132 |
| Solubility Information | Soluble in clear yellow solution 5% in ethanol,dimethyl sulfoxide and methanol. |
| Formula Weight | 161.16 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Indole-2-carboxylic acid |
Safety and Handling
| Hazard Category | H302+H312-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H311-H315-H319-H335 Toxic if swallowed. Toxic in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 216-030-4 |
| RTECSNumber | NK7882812 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.