Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-3-acetamide 98.0+%, TCI America™
SDP

Catalog No. i06685g
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 879-37-8
Molecular Formula C10H10N2O
Molecular Weight (g/mol) 174.20
MDL Number MFCD00005641
InChI Key ZOAMBXDOGPRZLP-UHFFFAOYSA-N
PubChem CID 397
ChEBI CHEBI:16031
IUPAC Name 2-(1H-indol-3-yl)acetamide
SMILES NC(=O)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 5 g
Formula Weight 174.20
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Indole-3-acetamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.