Learn More
Indole-3-acetic acid, 98+%
Description
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.
Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼167°C (decomposition) |
| Flash Point | 171°C (339°F) |
| Quantity | 25 g |
| Beilstein | 143358 |
| Sensitivity | Light sensitive |
| Merck Index | 14,4964 |
| Solubility Information | Soluble in ethanol (50mg/ml),methanol,DMSO,and chloroform (sparingly). Insoluble in water. |
| Formula Weight | 175.19 |
| Percent Purity | ≥98% |
| Chemical Name or Material | Indole-3-acetic acid |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 201-748-2 |
| RTECSNumber | NL3150000 |
| TSCA | Yes |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.