Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-3-butyric acid, 98%, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
100g
133-32-4
C12H13NO2
203.241
MFCD00005664
JTEDVYBZBROSJT-UHFFFAOYSA-N
indole-3-butyric acid,3-indolebutyric acid,4-1h-indol-3-yl butanoic acid,indolebutyric acid,hormodin,1h-indole-3-butanoic acid,seradix,indole-3-butanoic acid,jiffy grow,4-indol-3-yl butyric acid
8617
CHEBI:33070
4-(1H-indol-3-yl)butanoic acid
C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O
133-32-4
C12H13NO2
203.241
MFCD00005664
JTEDVYBZBROSJT-UHFFFAOYSA-N
indole-3-butyric acid,3-indolebutyric acid,4-1h-indol-3-yl butanoic acid,indolebutyric acid,hormodin,1h-indole-3-butanoic acid,seradix,indole-3-butanoic acid,jiffy grow,4-indol-3-yl butyric acid
8617
CHEBI:33070
4-(1H-indol-3-yl)butanoic acid
C1=CC=C2C(=C1)C(=CN2)CCCC(=O)O
Applications
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.

Solubility
Soluble in water(0.25g/L).

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Melting Point 121°C to 125°C
Quantity 25g
Odor Odorless
Assay Percent Range 98%
UN Number UN2811
Beilstein 171120
Merck Index 14,4965
Solubility Information Soluble in water(0.25g/L).
Formula Weight 203.24
Percent Purity 98%
Chemical Name or Material Indole-3-butyric acid
Show More Show Less

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 205-101-5

RTECSNumber : NL5250000

TSCA : Yes

storageNote1 : Air sensitive

Recommended Storage : Ambient temperatures