Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-3-Butyric Acid Potassium Salt, MP Biomedicals™

Catalog No. ICN10204325 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
25 g

Synonym: K-IBA, Potassium 4-(1H-Indol-3-yl)butanoate, 1H-Indole-3-Butanoic Acid Potassium Salt

Water-soluble form of IBA, an auxin-family plant growth hormone.

  • Promotes root organogenesis
  • Induces callus formation
  • Forms adventitious roots
  • Aids in regulation of gravitropism and phototropism
TRUSTED_SUSTAINABILITY

Specifications

CAS 60090-23-3
Quantity 25 g
Physical Form Powder

Unless specified otherwise, MP Biomedical™'s products are for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.