Learn More
Specifications
Specifications
| Concentration | 1 mg/mL |
| For Use With (Application) | Western Blot, ELISA |
| Formulation | PBS (pH 7.4) |
| Quantity | 0.1 mg |
| Storage Requirements | Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. |
| Purity or Quality Grade | Chromatography |
| Protein | Indole-3-butyric acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
