Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indoleamine 2,3-dioxygenase/IDO Antibody (700838) [DyLight 680], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6030Y
Description
Indoleamine 2,3-dioxygenase/IDO Monoclonal antibody specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
Indoleamine 2,3-dioxygenase/IDO | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
700838 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase | |
E. coli-derived recombinant human Indoleamine 2,3-dioxygenase/IDO, Ala2-Gly403, Accession # P14902 | |
0.1 mL | |
Cancer, Myeloid derived Suppressor Cell | |
3620 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction