Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indoleamine 2,3-dioxygenase/IDO Antibody (9E5), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Indoleamine 2,3-dioxygenase/IDO |
---|---|
Clone | 9E5 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Indoleamine 2,3-dioxygenase/IDO Monoclonal antibody specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Indoleamine 2,3-dioxygenase/IDO | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase | |
A synthesized peptide derived from Human Indoleamine 2,3-dioxygenase/IDO [UniProt P14902] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9E5 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Myeloid derived Suppressor Cell | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
3620 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title