Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indoleamine 2,3-dioxygenase/IDO Antibody (BLR031F), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP276404

Catalog No. NBP276404


Only null left
Add to Cart

Description

Description

Indoleamine 2,3-dioxygenase/IDO Monoclonal specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

Indoleamine 2,3-dioxygenase/IDO
Monoclonal
Unconjugated
Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA with 0.09% Sodium Azide
IDO1
Recombinant protein representing Human IDO1
100 μL
3620.0
Store at 2-8°C/ 1 year from date of receipt
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
BLR031F
Western Blot 1:1000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 -1:500, Immunohistochemistry-Paraffin
EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase
Rabbit
Antigen Affinity-purified
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.