Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indoleamine 2,3-dioxygenase/IDO Antibody (SR1771), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Indoleamine 2,3-dioxygenase/IDO |
---|---|
Clone | SR1771 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
Indoleamine 2,3-dioxygenase/IDO Monoclonal antibody specifically detects Indoleamine 2,3-dioxygenase/IDO in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Indoleamine 2,3-dioxygenase/IDO | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 1.13.11.52, IDOIDO-1, INDOindole 2,3-dioxygenase, indoleamine 2,3-dioxygenase 1, indoleamine-pyrrole 2,3 dioxygenase, Indoleamine-pyrrole 2,3-dioxygenase | |
A synthesized peptide derived from human Indoleamine 2,3-dioxygenase/IDO (Uniprot #: P14902) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1771 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Myeloid derived Suppressor Cell | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
3620 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title