Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(+/-)-Indoline-2-carboxylic acid, 95%
Click to view available options
Quantity:
5 g
25 g
Description
(±)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(+/-)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
(+/-)-Indoline-2-carboxylic acid is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼168°C (decomposition) |
| Quantity | 5 g |
| Solubility Information | Insoluble in water. |
| Formula Weight | 163.18 |
| Percent Purity | 95% |
| Chemical Name or Material | (+/-)-Indoline-2-carboxylic acid |
Safety and Handling
| Hazard Category | H317-H361 |
| Hazard Statement | GHS H Statement H361-H373-H317 Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. May cause an allergic skin reaction. |
| Precautionary Statement | P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c |
| EINECSNumber | 278-899-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction