Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium tin oxide coated glass slide, rectangular

Catalog No. 63691625PAK Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 pak
25 pak

Catalog No. 63-691-625PAK

Supplier: Sigma Aldrich 63691625PAK

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Indium tin oxide (ITO) is widely used as transparent conducting oxides. Its major characteristics are electrical conductivity, optical transparency and the ease to be deposited as films. ITO coated glass has a sheet resistance of16+¼/cm2.ITO coated glass slides were used as substrates for barium vanadate films,α- andα-Fe2O3 nanostructures. The product was used in a study of photoelectrical properties of spin coated polyphenylene films. Poly(N-vinylcarbazole) was spin coated onto the substrate to form a light emitting layer towards the fabrication of light emitting diode (LED).

Specifications

CAS 50926-11-9
Refractive Index n20/D 1.517 (lit.)
Linear Formula In2O3 -+ (SnO2)x
MDL Number MFCD00171662
Quantity 25 pak
Synonym ITO; ITO coated slide, rectangular
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.