Learn More
Industrial Scientific 58/103L Calibration Regulator
Provide proper flow rate for calibrating gas detectors.
Supplier:  Industrial Scientific 18101766
Catalog No. 19-822-268
Only null left
Add to Cart
Description
Description
  • For 58/103L calibration cylinders
  • Flow Rate: 1L/min.
Specifications
Specifications
58/103 L Calibration Cylinders
Regulator
Provide proper flow rate for calibrating gas detectors.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.