Learn More
BINDER™ Inert Gas Connection
SDP
Catalog No. 01672902 Shop All BINDER Inc. Products
Click to view available options
For Use With (Equipment):
Units M 115
Units M 260
Units M 56
Inert gas connection with gas inlet and outlet. Includes additional measures for the greatest possible impermeability to gas. Not in conjunction with access port or interior lighting option.
  • Inert Gas Connection
  • For use with various BINDER™ Unit M lines
Specifications
Volume (Metric) Gas None
For Use With (Equipment) Units M 56
Weight (English) 2.20 lb.
Weight (Metric) 1 kg
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.