Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BINDER™ Inert Gas Connection
SDP

Inert gas connection with gas inlet and outlet. Includes additional measures for the greatest possible impermeability to gas. Not in conjunction with access port or interior lighting option.

$1786.45

Specifications

Volume (Metric) Gas None
Weight (English) 2.20 lb.
Weight (Metric) 1 kg
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
01-672-900
SDP
View Documents
BINDER™
80122317
Units M 260
Each for $1,786.45
Only null left
Add to Cart
 
01-672-901
SDP
View Documents
BINDER™
80122316
Units M 115
Each for $1,786.45
Only null left
Add to Cart
 
01-672-902
SDP
View Documents
BINDER™
80122315
Units M 56
Each for $1,786.45
Only null left
Add to Cart
 
Description

Description

  • Inert Gas Connection
  • For use with various BINDER™ Unit M lines
Specifications

Specifications

None
2.20 lb.
1 kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.