Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ INF2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578560 Supplier Invitrogen™ Supplier No. PA578560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (90%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene represents a member of the formin family of proteins. It is considered a diaphanous formin due to the presence of a diaphanous inhibitory domain located at the N-terminus of the encoded protein. Studies of a similar mouse protein indicate that the protein encoded by this locus may function in polymerization and depolymerization of actin filaments. Mutations at this locus have been associated with focal segmental glomerulosclerosis 5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen INF2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene INF2
Gene Accession No. Q27J81
Gene Alias 2610204M08Rik; AA589465; AW125550; C14orf151; C14orf173; CMTDIE; EG629699; formin, inverted; FSGS5; HBEAG-binding protein 2 binding protein C; HBEBP2-binding protein C; Inf2; inverted formin; inverted formin 2; inverted formin, FH2 and WH2 domain containing; inverted formin-2; MGC13251; pp9484
Gene Symbols INF2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human INF2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.