Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ INF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5114519
Description
Positive controls that may be used include: A431 Cell Lysate.
This gene represents a member of the formin family of proteins. It is considered a diaphanous formin due to the presence of a diaphanous inhibitory domain located at the N-terminus of the encoded protein. Studies of a similar mouse protein indicate that the protein encoded by this locus may function in polymerization and depolymerization of actin filaments. Mutations at this locus have been associated with focal segmental glomerulosclerosis 5.
Specifications
INF2 | |
Polyclonal | |
Unconjugated | |
INF2 | |
2610204M08Rik; AA589465; AW125550; C14orf151; C14orf173; CMTDIE; EG629699; formin, inverted; FSGS5; HBEAG-binding protein 2 binding protein C; HBEBP2-binding protein C; Inf2; inverted formin; inverted formin 2; inverted formin, FH2 and WH2 domain containing; inverted formin-2; MGC13251; pp9484 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64423 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q27J81 | |
INF2 | |
INF2 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human INF2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction