Learn More
INF2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303428AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene represents a member of the formin family of proteins. It is considered a diaphanous formin due to the presence of a diaphanous inhibitory domain located at the N-terminus of the encoded protein. Studies of a similar mouse protein indicate that the protein encoded by this locus may function in polymerization and depolymerization of actin filaments. Mutations at this locus have been associated with focal segmental glomerulosclerosis 5.Specifications
INF2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q27J81 | |
INF2 | |
Between 950 and 1000 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
INF2 | |
C14orf151, C14orf173, chromosome 14 open reading frame 151, chromosome 14 open reading frame 173, CMTDIE, FSGS5, HBEAG-binding protein 2 binding protein C, HBEBP2-binding protein C, inverted formin 2, MGC13251, pp9484 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
64423 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.