Description
- Sony Pregius global shutter sensor technology
- Color IMX252 Global Shutter CMOS sensor with 1/1.8 in. optical format providing a resolution of 2064 x 1544 using 3.45 x 3.45μm pixels
- Fast frame rates at approximately 120 fps at full resolution in 8-bit
- Dual output to USB 3.0 and HDMI compatible monitor
- Intuitive on-camera buttons for power, white balance, and image capture
- Selectable 8 or 12-bit pixel data
- High-speed USB 3.1 Gen 1 interface for fastest image delivery and simplified connectivity. USB 2.0 supported.
- Software compatible with Windows 10, Windows 8.1, Windows 7, MAC OS X 10.13, 32 and 64-bit operating systems. Includes TWAIN and DirectShow interfaces.
- Support for third-party capture and analysis applications: MetaMorph and Micro-Manager
- 0.5x or 0.63x camera coupler recommended
Specifications
Specifications
| Type | USB Microscope Camera |
| Autoclavable | Not Autoclavable |
| Sterility | Non-sterile |
| Voltage | 5 V |