Learn More
ACCU-SCOPE™ INFINITY 5 MP Camera
SDP
Catalog No. 01189217 Shop All ACCU-SCOPE Products
Change view
Click to view available options
Voltage:
5 VDC
Catalog No. Voltage
01-189-217 5 VDC
1 options
Catalog No. 01-189-217 Supplier ACCU-SCOPE™ Supplier No. AU510CCMOS
Only null left
Add to Cart
Add to Cart
Request a Quote
The INFINITY 5-5C is a high quality 5-megapixel microscopy camera that provides high speeds at high resolution. Based on the Sony™ Pregius™ global shutter CMOS sensor. The INFINITY 5 is ready to perform with cables (USB and HDMI), power supply, and INFINITY CAPTURE software for routine imaging needs
  • Sony Pregius global shutter sensor technology
  • Color IMX252 Global Shutter CMOS sensor with 2/3 in. optical format providing a resolution of 2464 x 2056 using 3.45 x 3.45μm pixels
  • Fast frame rates at approximately 75 fps at full resolution in 8-bit
  • Dual output to USB 3.0 and HDMI compatible monitor
  • Intuitive on-camera buttons for power, white balance and image capture
  • Selectable 8 or 12-bit pixel data
  • High-speed USB 3.1 Gen 1 interface for fastest image delivery and simplified connectivity. USB 2.0 supported.
  • Software compatible with Windows 10, Windows 8.1, Windows 7, MAC OS X 10.13, 32 and 64-bit operating systems. Includes TWAIN and DirectShow interfaces.
  • Support for third-party capture and analysis applications: MetaMorph and Micro-Manager
  • 0.6x to 0.7x camera coupler recommended
Specifications
Type USB Microscope Camera
Autoclavable Not Autoclavable
Sterility Non-sterile
Voltage 5 VDC