Learn More
Motic™ Swift Line Infinity Compound Microscope, Binocular Head
SCI_ED
Catalog No. s81766 Shop All Motic Swift Line Products
Click to view available options
Objective Types:
Infinity Corrected
Catalog No. S81766
Supplier: Motic™ Swift Line M17BP
Only null left
Add to Cart
Add to Cart
The M17 microscope series features infinity corrected plan optics ideal for college laboratories veterinary and medical applications. Supplied with binocular head this research level microscope also incorporates 3W LED illumination.
The M17 compound microscope series features infinity corrected plan optics, ideal for college laboratories, veterinary and medical applications. Supplied with binocular head, this research level microscope also incorporates 3W LED illumination and wide field 20 mm eyepieces for a very crisp image.
OPTICAL SYSTEM
  • Widefield 10x /20 mm eyepieces
  • Binocular viewing head with 30° inclined eyepieces, with diopter and interpupillary adjustment 54 mm to 76 mm
  • Infinity corrected 4x (0.10 N.A.), 10x (0.25 N.A.), 40xR (0.65 N.A.) and 100xR oil immersion (1.25 N.A.) objectives. (Only the 100xR objective is oil immersion.) PLAN lenses. All are parfocalled, parcentered and color-coded
  • Quad objective turret is ball-bearing mounted for smooth, precise positioning of objectives. Mounted in reverse position to facilitate ease in changing slides
ILLUMINATION
  • 3W LED
  • Illumination on/off switch
ELECTRICAL
  • 110 V to 220 V switching power supply, unit shipped with American plug
FOCUSING
  • Coaxial coarse and fine focusing controls, coarse has tension adjustment, fine is graduated
STAGE
  • Large 5-3/4 in. x 5-1/2 in. (142 mm x 140 mm) stage; built-in, low profile, ball-bearing mounted mechanical specimen holder with low-position coaxial controls
  • 1.25 N.A. Abbe condenser has rack & pinion focusing and iris diaphragm
Specifications
Eyepiece WF10X/20 mm
Head Style Binocular
Lighting Type LED
Magnification Power 4X, 10X, 40XR, 100XR
Model M17B-P
Objective Types Infinity Corrected
Product Type Advanced Infinity Compound
Includes Vinyl Dustcover
Material Metal, Aluminum
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.