Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Infliximab Chimeric Recombinant Human Monoclonal Antibody (cA2 (Infliximab))
GREENER_CHOICE

Catalog No. PIMA547798
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547798 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547798 Supplier Invitrogen™ Supplier No. MA547798
Add to Cart
Add to Cart

Human Recombinant Monoclonal Antibody

Specificity: Binds to soluble and transmembrane forms of TNF-alpha and neutralizes its biological activity. Does not bind to TNF-beta.

Infliximab with the brand name of Remicade, is a chimeric monoclonal IgG1 antibody. It binds to TNF-alpha, which is a key proinflammatory cytokine involved in chronic inflammatory diseases. Infliximab functions as TNF-alpha blocker to disrupt the interaction of TNF-alpha with its receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Infliximab Chimeric
Applications ELISA, Flow Cytometry, Functional Assay, Western Blot
Classification Recombinant Monoclonal
Clone cA2 (Infliximab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene ABCB1
Gene Accession No. P08183
Gene Alias Remicade; TA-650
Host Species Human
Immunogen Purified recombinant human TNF (rTNF).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5243
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.