Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZeptoMetrix Influenza A H1 Virus Culture Fluid

Click to view available options
Strain Designations:
A/Brisbane/59/07, A/New Caledonia/20/99, A/PR/8/34, A/Taiwan/42/06
A/Solomon Islands/03/06
Description
IMPORTANT: Contains live material. Customer must submit an executed ZeptoMetrix MTA before vendor can ship order.
Specifications
Specifications
Biosafety Level | 2 |
Certifications/Compliance | ISO, 13485 |
Content And Storage | -65°C |
Form | Solution |
Media Type | Vial |
Product Type | Influenza A H1N1 Virus Infectious Culture Fluid |
For Use With (Application) | Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays |
Quantity | 1 mL |
Strain Designations | A/Brisbane/59/07, A/New Caledonia/20/99, A/PR/8/34, A/Taiwan/42/06 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction