Learn More
Invitrogen™ Influenza A H10N3 HA (A/Jiangsu/428/2021) Recombinant Rabbit Monoclonal Antibody (HL1559)

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | Influenza A H10N3 HA (A/Jiangsu/428/2021) |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | HL1559 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | flu hemagglutinin; influenza hemagglutinin |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of Influenza A virus H10N3 HA (Hemagglutinin)(A/Jiangsu/428/2021 (H10N3)). The exact sequence is proprietary. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.