Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H1N1/H3N2 M1 Antibody (GA2B), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10066552AF350
Description
Influenza A H1N1/H3N2 M1 Monoclonal antibody specifically detects Influenza A H1N1/H3N2 M1 in Virus samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Influenza A H1N1/H3N2 M1 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Influenza A/ Puerto Rico/ 8/ 34 (H1N1) and A/Bangkok/ 1/ 79 (H3N2) viruses. | |
0.5 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
GA2B | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction