Learn More
Invitrogen™ Influenza A H1N1 HA (A/New Caledonia/20/99) Polyclonal Antibody

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Influenza A H1N1 (A/New Caledonia/20/99) Hemagglutinin/HA.
Specifications
Specifications
| Antigen | Influenza A H1N1 HA (A/New Caledonia/20/99) |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; Influenza H1N1; influenza hemagglutinin |
| Host Species | Rabbit |
| Immunogen | Recombinant Influenza A H1N1 (A/New Caledonia/20/99) Hemagglutinin/HA Protein. |
| Purification Method | Protein A |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.