Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Influenza A H1N1 PB2 (A/Puerto Rico/8/34) Recombinant Rabbit Monoclonal Antibody (HL1994)
GREENER_CHOICE

Catalog No. FEMA550357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550357 Supplier Invitrogen™ Supplier No. MA550357
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Influenza A H1N1 PB2 (A/Puerto Rico/8/34) Recombinant Monoclonal Antibody for Western Blot, ICC/IF

Influenza A (H1N1) virus is a subtype of influenza virus A and is the most common cause of influenza (flu) in humans. A strain of swine-origin H1N1 was responsible for the 2009 flu pandemic. H1N1PB2 protein is a well-documented component of the viral polymerase complex required for virus replication. The PB2 protein is involved in host range restriction and pathogenicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Influenza A H1N1 PB2 (A/Puerto Rico/8/34)
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1994
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PB2
Gene Accession No. P03428
Gene Alias flu hemagglutinin, H1N1 HA, H1N1 hemagglutinin, Influenza H1N1, influenza hemagglutinin
Gene Symbols PB2
Host Species Rabbit
Immunogen Recombinant fragmemt of Influenza A virus PB2 protein.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 956536
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.