Learn More
Invitrogen™ Influenza A H1N1 HA Polyclonal Antibody

Description
This antibody detects a peptide sequence that differs from Product # PA5-20713 and PA5-20715, and can be used as a cognate pair with Product # PA5-20731. PA5-20712 can be used with blocking peptide PEP-0827.
Specifications
Specifications
| Antigen | Influenza A H1N1 HA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | flu hemagglutinin; H1N1 HA; H1N1 hemagglutinin; Influenza H1N1; influenza hemagglutinin |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from the Hemagglutinin protein. |
| Purification Method | Antigen affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.