Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H1N1 Hemagglutinin Antibody (10), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP306578R
Description
Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza virus N1 samples. It is validated for Western BlotSpecifications
Influenza A H1N1 Hemagglutinin | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
This antibody was obtained from a rabbit immunized with purified, recombinant Influenza H1N1 (A/California/04/2009) Hemagglutinin. | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
10 | |
Western Blot | |
Rabbit | |
Protein A purified | |
RUO | |
Influenza virus N1 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction