Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H1N1 Hemagglutinin Antibody (2F1A7), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306443G
Description
Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Influenza virus N1 samples. It is validated for Western Blot, ELISA, ELISA, ELISASpecifications
Influenza A H1N1 Hemagglutinin | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant H1N1 (A/Puerto Rico/8/1934) HA (ABD77675.1; Met 1-Gln 528). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
2F1A7 | |
Western Blot, ELISA, Sandwich ELISA, Sandwich ELISA Detection | |
Mouse | |
Protein A purified | |
RUO | |
Influenza virus N1 | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction