Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H1N1 Hemagglutinin Antibody (9G1G8), Novus Biologicals™

Mouse Monoclonal Antibody
$170.70 - $463.50
Specifications
Antigen | Influenza A H1N1 Hemagglutinin |
---|---|
Clone | 9G1G8 |
Dilution | Western Blot 1:500 - 1:1000, ELISA 1:1000-1:2000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
Influenza A H1N1 Hemagglutinin Monoclonal antibody specifically detects Influenza A H1N1 Hemagglutinin in Virus samples. It is validated for Western Blot, ELISASpecifications
Influenza A H1N1 Hemagglutinin | |
Western Blot 1:500 - 1:1000, ELISA 1:1000-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
IgG2b | |
Protein A purified |
9G1G8 | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Virus | |
Recombinant Influenza A H1N1 Hemagglutinin | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title