Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Hemagglutinin Antibody (IV.C102) - (A/USSR/90/77) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199821 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017687 0.025 mg
NB10065047 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017687 Supplier Novus Biologicals Supplier No. NB100650470.025MG

Mouse Monoclonal Antibody

Influenza A Haemagglutinin H1N1 Monoclonal specifically detects Influenza A Haemagglutinin H1N1 in Influenza A Virus H1N1, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Sandwich ELISA.

Specifications

Antigen Influenza A Haemagglutinin H1N1
Applications Immunofluorescence, ELISA, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry
Classification Monoclonal
Clone IV.C102
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation PBS (pH 7.2) with 0.09% Sodium Azide
Gene Accession No. P03453
Gene Alias HA, HA1, HA2, Hemagglutinin, Influenza A Virus Hemagglutinin
Gene Symbols HA
Host Species Mouse
Immunogen Influenza A/USSR/90/77
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-65047 is specific for the haemagglutinin of the Influenza A H1N1 serotype and does not react with H2N2, H3N2, Influenza B, RSV, Parainfluenza I, II or III, Adenovirus, mumps or measle antigens.
Target Species Virus, Influenza virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.