Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Influenza A H1N1 M1 Antibody (02), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306633MFV500
Description
Influenza A H1N1 M1 Monoclonal antibody specifically detects Influenza A H1N1 M1 in Influenza virusN1 samples. It is validated for ELISASpecifications
| Influenza A H1N1 M1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Influenza A H1N1 (Accession#: NP_040978.1; Ser 2-Lys 252). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| ELISA | |
| 02 | |
| ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Influenza virus N1 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction