Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 M1 Antibody - (A/Puerto Rico/8/1934), Novus Biologicals™
SDP

Catalog No. NBP214996 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214996 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP214996 Supplier Novus Biologicals Supplier No. NBP214996
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Influenza A H1N1 M1 Polyclonal antibody specifically detects Influenza A H1N1 M1 in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Influenza A H1N1 M1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA
Formulation 1XPBS, 1% BSA, 20% Glycerol (pH7). with 0.025% Proclin 300
Gene Accession No. H1N1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Internal region of Influenza A virus M1 (matrix protein) (A/Puerto Rico/8/1934(H1N1). The exact sequence is proprietary.
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Influenza A Virus.
Target Species Virus
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.