Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A H1N1 Neuraminidase Antibody - (A/WSN/1933), Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP242870

Catalog No. NBP242870


Only null left
Add to Cart

Description

Description

Specifically detects Influenza A Virus H1N1 Neuraminidase in Virus samples, and it is validated for Western Blotting

Influenza A H1N1 Neuraminidase Polyclonal antibody specifically detects Influenza A H1N1 Neuraminidase in Influenza A Virus H1N1 samples. It is validated for Western Blot.
Specifications

Specifications

Influenza A H1N1 Neuraminidase
Polyclonal
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence
Rabbit
50 kDa
0.1 mL
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Immunofluorescence
Unconjugated
PBS, 1% BSA, 20% Glycerol (pH7) with 0.025% Proclin 300
Recombinant protein encompassing a sequence within the N-terminus region of Influenza A virus H1N1 NA (Neuraminidase) (A/WSN/1933(H1N1). The exact sequence is proprietary.
Affinity Purified
RUO
Influenza A virus H1N1, Influenza virus
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only