Learn More
Invitrogen™ Influenza A H1N1 NP Monoclonal Antibody (F8)

Description
MA1-10512 detects Influenza A Virus Hemagglutinin. This antibody recognizes all serotypes of Influenza A virus and inhibits viral replication after fatty acid acylation. MA1-10512 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10512 immunogen is Influenza virus type A strain H1N1.
Specifications
Specifications
| Antigen | Influenza A H1N1 NP |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | F8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene Alias | H1N1 HA; H1N1 hemagglutinin; Influenza A; Influenza H1N1 |
| Host Species | Mouse |
| Immunogen | Influenza virus type A strain H1N1. |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.